Anti-CES1 Rabbit Polyclonal Antibody
Catalog # ABNOH00001066-D01P
Supplier: Abnova
Specifications
- Antibody type:Primary
- Antigen name:Carboxylesterase 1
- Antigen symbol:CES1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Reactivity:Human
- Western blot:Yes
- Cross adsorption:No
- Gene ID:1066
- Antigen synonyms:CES2|CEH|carboxylesterase 2 (liver)|HMSE|carboxylesterase 1|SES1|triacylglycerol hydrolase|cholesteryl ester hydrolase|CES1|monocyte/macrophage serine-esterase 1|liver carboxylesterase|carboxylesterase 1 (monocyte/macrophage serine esterase 1)|MGC117365|ACAT|HMSE1|egasyn|acyl coenzyme A:cholesterol acyltransferase|TGH|PCE-1
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:100 µg
- Pk:100 µg
Specifications
About this item
Type: Primary
Antigen: CES1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human