Anti-CDSN Mouse Monoclonal Antibody [clone: 5B4]
Katalogové číslo ABNOH00001041-M02
Dodavatel: Abnova
Specifikace
- Antibody type:Primary
- Antigen name:Corneodesmosin
- Antigen symbol:CDSN
- Clonality:Monoclonal
- Clone:5B4
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- Reactivity:Human
- Western blot:Yes
- Cross adsorption:No
- Gene ID:1041
- Antigen synonyms:HYPT2|HTSS|S|PSS1|PSS|HTSS1
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:100 µg
- Pk:100 µg
Specifikace
O tomto produktu
Type: Primary
Antigen: CDSN
Clonality: Monoclonal
Clone: 5B4
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human