Anti-CDSN Mouse Monoclonal Antibody [clone: 6F11]
Catalog # ABNOH00001041-M01
Supplier: Abnova
Specifications
- Antibody type:Primary
- Antigen name:Corneodesmosin
- Antigen symbol:CDSN
- Clonality:Monoclonal
- Clone:6F11
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- Reactivity:Human
- Western blot:Yes
- Cross adsorption:No
- Gene ID:1041
- Antigen synonyms:HYPT2|HTSS|S|PSS1|PSS|HTSS1
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:100 µg
- Pk:100 µg
Specifications
About this item
Type: Primary
Antigen: CDSN
Clonality: Monoclonal
Clone: 6F11
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human