Anti-SLC25A20 Mouse Monoclonal Antibody [clone: M2]
Catalog # ABNOH00000788-M02
Supplier: Abnova
Specifications
- Antibody type:Primary
- Antigen name:Solute Carrier Family 25 Member A20
- Antigen symbol:Solute carrier family 25, member 20
- Clonality:Monoclonal
- Clone:M2
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- ImmunoFluorescence:Yes
- Reactivity:Human
- Cross adsorption:No
- Gene ID:788
- Antigen synonyms:CACT|CAC|Solute carrier family 25 member 20|Carnitine/acylcarnitine translocase|MCAT_HUMAN.
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:100 µg
- Pk:100 µg
Specifications
About this item
Type: Primary
Antigen: SLC25A20
Clonality: Monoclonal
Clone: M2
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human