Anti-BCAT1 Mouse Monoclonal Antibody [clone: 1F8]
Catalog # ABNOH00000586-M02
Supplier: Abnova
Specifications
- Antibody type:Primary
- Antigen name:Branched Chain Amino-Acid Transaminase 1, Cytosolic
- Antigen symbol:BCAT1
- Clonality:Monoclonal
- Clone:1F8
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- Reactivity:Human
- Cross adsorption:No
- Gene ID:586
- Antigen synonyms:BCT1|PNAS121|ECA39|BCATC|PP18|MECA39
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:100 µg
- Pk:100 µg
Specifications
About this item
Type: Primary
Antigen: BCAT1
Clonality: Monoclonal
Clone: 1F8
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human