Anti-SELE Mouse Monoclonal Antibody [clone: OTICL-37]
Catalog # ORIGTA600028
Supplier: OriGene
Specifications
- Antibody type:Primary
- Antigen name:Selectin E
- Antigen symbol:SELE
- Clonality:Monoclonal
- Clone:OTICL-37
- Conjugation:Unconjugated
- Host:Mouse
- Isotype:IgG1
- Reactivity:Human
- Cross adsorption:none
- Form:Liquid
- Storage buffer:Stored in PBS SELE antibody clone CL-37 in aqueous buffer, no sodium azide
- Storage temperature:Cool
- Shipping temperature:Ice
- Pk:100 µG
Specifications
About this item
Recommended Dilutions: 1:100-1:1000
Type: Primary
Antigen: SELE (selectin E)
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human