Anti-PPAT Mouse Monoclonal Antibody [clone: OTI1B2]
ORIGTA504559
: OriGene
- Antibody type:Primary
- Antigen name:phosphoribosyl pyrophosphate amidotransferase
- Antigen symbol:PPAT
- Clonality:Monoclonal
- Clone:OTI1B2
- Conjugation:Unconjugated
- Flow cytometry:Yes
- Host:Mouse
- ImmunoFluorescence:Yes
- Isotype:IgG1
- Western blot:Yes
- Cross adsorption:none
- Form:Liquid
- Amino acid number:42 to 278
- Storage buffer:PBS (PH 7.3) containing 1% BSA, 50% glycerol and 0,02% sodium azide.
- Molecular weight:57.2 kDa
- Storage temperature:Cool
- Shipping temperature:Ice
- Immunogen:Protein
- Purification:purified by affinity chromatography
- Pk:100 µl
Recommended Dilutions: Western Blot: 1:500-2000; Flow Cytometry: 1:100
Type: Primary
Antigen: PPAT (phosphoribosyl pyrophosphate amidotransferase)
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: