Order Entry
Czech Republic
ContactUsLinkComponent
Anti-SLC3A2 Mouse Monoclonal Antibody [clone: 44D7]
Anti-SLC3A2 Mouse Monoclonal Antibody [clone: 44D7]
  ORIGTA318205
 :  OriGene
Anti-SLC3A2 Mouse Monoclonal Antibody [clone: 44D7]
  ORIGTA318205
 :  OriGene

 

  • Antibody type:
    Primary
  • Antigen name:
    solute carrier family 3 (amino acid transporter heavy chain), member 2
  • Antigen symbol:
    Solute carrier family 3, member 2
  • Clonality:
    Monoclonal
  • Clone:
    44D7
  • Conjugation:
    Unconjugated
  • ELISA:
    Yes
  • Host:
    Mouse
  • ImmunoChemistry:
    Yes
  • Isotype:
    IgG1
  • Reactivity:
    Human
  • Western blot:
    Yes
  • Cross adsorption:
    none
  • Form:
    Liquid
  • Storage buffer:
    PBS, 0,09% sodium azide
  • Storage temperature:
    Cool
  • Shipping temperature:
    Ice
  • Purification:
    purified by Protein G chromatography
  • Pk:
    50 µG

 

 

Recommended Dilutions: Flo: 1:50-1:200; Immunochemistry: 15 µg/ml

Type: Primary
Antigen: SLC3A2 (solute carrier family 3 (amino acid transporter heavy chain), member 2)
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human