Order Entry
Czech Republic
ContactUsLinkComponent
Anti-CACNA1H Mouse Monoclonal Antibody [clone: S55-10]
Anti-CACNA1H Mouse Monoclonal Antibody [clone: S55-10]
  ORIGTA309309
 :  OriGene
Anti-CACNA1H Mouse Monoclonal Antibody [clone: S55-10]
  ORIGTA309309
 :  OriGene

 

  • Antibody type:
    Primary
  • Antigen name:
    calcium channel, voltage-dependent, T type, alpha 1H subunit
  • Antigen symbol:
    CACNA1H
  • Clonality:
    Monoclonal
  • Clone:
    S55-10
  • Conjugation:
    Unconjugated
  • ELISA:
    Yes
  • Flow cytometry:
    Yes
  • Host:
    Mouse
  • ImmunoChemistry:
    Yes
  • ImmunoPrecipitation:
    Yes
  • Isotype:
    IgG1
  • Reactivity:
    Human
  • Western blot:
    Yes
  • Cross adsorption:
    none
  • Form:
    Liquid
  • Storage buffer:
    PBS pH 7,4, 50% glycerol
  • Storage temperature:
    Cool
  • Shipping temperature:
    Ice
  • Pk:
    100 µG

 

 

Recommended Dilutions:

Type: Primary
Antigen: CACNA1H (calcium channel, voltage-dependent, T type, alpha 1H subunit)
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human