Anti-CLIC3 Rabbit Polyclonal Antibody
ORIGTA308675
: OriGene
- Antibody type:Primary
- Antigen name:chloride intracellular channel 3
- Antigen symbol:CLIC3
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG
- Reactivity:Human
- Western blot:Yes
- Cross adsorption:none
- Form:Liquid
- Amino acid number:1 to 236
- Storage buffer:0,1M Tris, 0,1M Glycine, 10% Glycerol (pH7). 0,01% Thimerosal was added as a preservative.
- Molecular weight:27 kDa
- Storage temperature:Cool
- Shipping temperature:Ice
- Immunogen:Protein
- Purification:purified by antigen affinity chromatography
- Pk:100 µl
Recommended Dilutions: Immunochemistry1:100-1:1000; Western Blot:1:500-1:3000
Type: Primary
Antigen: CLIC3 (chloride intracellular channel 3)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human