Order Entry
Czech Republic
ContactUsLinkComponent
Anti-RGPD5 Rabbit Polyclonal Antibody
Anti-RGPD5 Rabbit Polyclonal Antibody
Catalog # ORIGTA306566
Supplier:  OriGene
Anti-RGPD5 Rabbit Polyclonal Antibody
Catalog # ORIGTA306566
Supplier:  OriGene

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    RANBP2-like and GRIP domain containing 5
  • Antigen symbol:
    RGPD5
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • Host:
    Rabbit
  • ImmunoChemistry:
    Yes
  • Isotype:
    IgG
  • Reactivity:
    Human
  • Western blot:
    Yes
  • Cross adsorption:
    none
  • Form:
    Liquid
  • Storage buffer:
    PBS containing 0,02% sodium azide.
  • Storage temperature:
    Cool
  • Shipping temperature:
    Ice
  • Purification:
    purified by affinity chromatography via peptide column
  • Pk:
    100 µG

Specifications

About this item

Recommended Dilutions: Western Blot: 1 ?g/ml; Immunochemistry: 10 ?g/ml; Immunofluorescence: 20 ?g/ml

Type: Primary
Antigen: RGPD5 (RANBP2-like and GRIP domain containing 5)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human