Order Entry
Czech Republic
ContactUsLinkComponent
Anti-CES1 Goat polyclonal antibody
Anti-CES1 Goat polyclonal antibody
Katalogové číslo ORIGTA302448
Dodavatel:  OriGene
Anti-CES1 Goat polyclonal antibody
Katalogové číslo ORIGTA302448
Dodavatel:  OriGene

Specifikace

  • Antibody type:
    Primary
  • Antigen name:
    Carboxylesterase 1
  • Antigen symbol:
    CES1
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • ELISA:
    Yes
  • Host:
    Goat
  • Isotype:
    IgG
  • Reactivity:
    Human
  • Western blot:
    Yes
  • Epitope:
    near C-terminus
  • Cross adsorption:
    none
  • Form:
    Liquid
  • Storage buffer:
    Supplied at 0,5 mg/ml in Tris saline, 0,02% sodium azide, pH 7,3 with 0,5% BSA.
  • Molecular weight:
    66.02 kDa
  • Sequence:
    C-NTQAAQKLKDKE
  • Storage temperature:
    Cool
  • Shipping temperature:
    Ice
  • Purification:
    purified by ammonium sulphate precipitation followed by antigen affinity chromatography
  • Pk:
    100 µG

Specifikace

O tomto produktu

Recommended Dilutions: ELISA: 1:8000; Western Blot: 003-01µg/ml

Type: Primary
Antigen: CES1 (carboxylesterase 1)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Goat
Isotype: IgG (Goat)
Reactivity: Human