Order Entry
Czech Republic
ContactUsLinkComponent
Anti-CLIC4 Rabbit Polyclonal Antibody
Anti-CLIC4 Rabbit Polyclonal Antibody
  ORIGTA302056
 :  OriGene
Anti-CLIC4 Rabbit Polyclonal Antibody
  ORIGTA302056
 :  OriGene

 

  • Antibody type:
    Primary
  • Antigen name:
    chloride intracellular channel 4
  • Antigen symbol:
    CLIC4
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • ELISA:
    Yes
  • Host:
    Rabbit
  • ImmunoChemistry:
    Yes
  • Isotype:
    IgG
  • Reactivity:
    Human
  • Western blot:
    Yes
  • Cross adsorption:
    none
  • Form:
    Liquid
  • Storage buffer:
    Purified polyclonal antibody supplied in PBS with 0,09% (W/V) sodium azide.
  • Molecular weight:
    28.77 kDa
  • Storage temperature:
    Cool
  • Shipping temperature:
    Ice
  • Immunogen:
    Protein
  • Purification:
    purified by saturated ammonium sulfate precipitation followed by dialysis against PBS
  • Pk:
    100 µG

 

 

Recommended Dilutions: Western Blot: 1:1000; Immunochemistry: 1:10-50

Type: Primary
Antigen: CLIC4 (chloride intracellular channel 4)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG (Rabbit)
Reactivity: Human