Order Entry
Czech Republic
ContactUsLinkComponent
Anti-CHAF1B Mouse Monoclonal Antibody [clone: SS 53]
Anti-CHAF1B Mouse Monoclonal Antibody [clone: SS 53]
Catalog # ORIGTA301488
Supplier:  OriGene
Anti-CHAF1B Mouse Monoclonal Antibody [clone: SS 53]
Catalog # ORIGTA301488
Supplier:  OriGene

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    chromatin assembly factor 1, subunit B (p60)
  • Antigen symbol:
    CHAF1B
  • Clonality:
    Monoclonal
  • Clone:
    SS 53
  • Conjugation:
    Unconjugated
  • Host:
    Mouse
  • ImmunoFluorescence:
    Yes
  • Isotype:
    IgG1
  • Reactivity:
    Human
  • Western blot:
    Yes
  • Cross adsorption:
    none
  • Format:
    Ascites
  • Form:
    Liquid
  • Storage buffer:
    Ascitic fluid and 0,1% sodium azide
  • Storage temperature:
    Cool
  • Shipping temperature:
    Ice
  • Immunogen:
    Protein
  • Pk:
    100 µl

Specifications

About this item

Recommended Dilutions: Western Blot: 1: 1000

Type: Primary
Antigen: CHAF1B (chromatin assembly factor 1, subunit B (p60))
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human