Specifications
- Antibody type:Primary
- Antigen name:MHC Class 2
- Antigen symbol:MHC2
- Clonality:Monoclonal
- Clone:M5/115.15.2
- Conjugation:Unconjugated
- Flow cytometry:Yes
- Host:Rat
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- ImmunoPrecipitation:Yes
- Isotype:IgG2b kappa
- Reactivity:Mouse
- Western blot:Yes
- Cross adsorption:none
- Gene ID:14961/14969
- Antigen synonyms:MHC II|IA/IE|IA|IE|I-A/E
- Storage temperature:Cool
- Shipping temperature:Ice
- Pk:100 µG
Specifications
About this item
The M5/114.15.2 antibody reacts with mouse MHC Class II alloantigens I-Ab, I-Ad, I-Aq, I-Ed, and I-Ek, as well as being cross-reactive with mouse cells of H-2p and H-2r haplotype. MHC Class II is widely expressed by mouse immune cells bearing these alloantigens, including T and B cells, monocytes, macrophages and dendritic cells. The antibody does not react with the following alloantigens: I-Af, I-Ak, I-As, or NOD H-2g7.
- Purified
The M5/114.15.2 antibody may be used for analysis of mouse cells expressing MHC Class II alloantigens as described. Please note that the M5/114.15.2 clone may also be referred to as M5/114 in the literature.
Type: Primary
Antigen: MHC II (I-A/I-E) (MHC II (I-A/I-E))
Clonality: monoclonal
Clone: M5/115.15.2
Conjugation: unconjugated
Epitope:
Host: Rat
Isotype: IgG2b Kappa
Reactivity: Mouse