Anti-CTLA4 Armenian Hamster monoclonal antibody [clone: UC10-4F10-11]
Catalog # 70-1522-U100
Supplier: Tonbo Biosciences
Specifications
- Antibody type:Primary
- Antigen name:Cytotoxic T-lymphocyte-Associated Protein 4
- Antigen symbol:CTLA4
- Clonality:Monoclonal
- Clone:UC10-4F10-11
- Conjugation:Unconjugated
- Flow cytometry:Yes
- Host:Armenian hamster
- Isotype:IgG1 kappa
- Reactivity:Mouse
- Cross adsorption:none
- Gene ID:12477
- Antigen synonyms:IDDM12|GRD4|GSE|CD|CTLA-4|ALPS5|cytotoxic T-lymphocyte associated protein 4|CD152|CELIAC3
- Storage temperature:Cool
- Shipping temperature:Ice
- Pk:100 µG
Specifications
About this item
Type: Primary
Antigen: CTLA4 (cytotoxic T-lymphocyte-associated protein 4)
Clonality: monoclonal
Clone: UC10-4F10-11
Conjugation: unconjugated
Epitope:
Host: Armenian Hamster
Isotype: IgG1 Kappa
Reactivity: Mouse