Order Entry
Czech Republic
ContactUsLinkComponent
Anti-CD274 Rat Monoclonal Antibody [clone: 10F.9G2]
Anti-CD274 Rat Monoclonal Antibody [clone: 10F.9G2]
Catalog # 70-1243-U100
Supplier:  Tonbo Biosciences
undefined
Anti-CD274 Rat Monoclonal Antibody [clone: 10F.9G2]
Catalog # 70-1243-U100
Supplier:  Tonbo Biosciences

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    CD274 molecule
  • Antigen symbol:
    CD274
  • Clonality:
    Monoclonal
  • Clone:
    10F.9G2
  • Conjugation:
    Unconjugated
  • Flow cytometry:
    Yes
  • Host:
    Rat
  • Isotype:
    IgG2b kappa
  • Reactivity:
    Mouse
  • Cross adsorption:
    none
  • Gene ID:
    60533
  • Antigen synonyms:
    PDL1|PD-L1|B7H1|B7-H|PDCD1L1|PDCD1LG1
  • Storage temperature:
    Cool
  • Shipping temperature:
    Ice
  • Pk:
    100 µG

Specifications

About this item

The 10F.9G2 antibody is specific for mouse CD274, more commonly known as PD-L1 or B7-H1, which acts as a ligand for the T cell co-regulatory receptor PD-1 (CD279). This interaction modulates T cell antigen receptor (TCR) signaling and therefore T cell activation. PD-L1 binding to PD-1 expressed on CD4- CD8- thymocytes participates in the processes of clonal selection, elimination of autoreactive lymphocytes, and development of tolerance. PD-L1 may also bind PD-1 following the receptor’s inducible expression on activated, mature T cells, where it has been proposed to limit T cell activation. PD-L1 is one of a group of “B7” ligands whose interactions with the CD28 receptor family, also including CTLA-4 (CD152), provide a balance of co-stimulatory /co-inhibitory signaling important in T cell activation, tolerance, and autoimmunity.

The 10F.9G2 antibody may be used as a marker for PD-L1 expression on T and B cells, NK cells and on dendritic cells. It is also widely used for analysis of receptor-ligand interaction and function(s) in vitro and in vivo.

Type: Primary
Antigen: CD274 (CD274 molecule)
Clonality: monoclonal
Clone: 10F.9G2
Conjugation: unconjugated
Epitope:
Host: Rat
Isotype: IgG2b Kappa
Reactivity: Mouse