Order Entry
Czech Republic
ContactUsLinkComponent
Anti-IL2RA Mouse Monoclonal Antibody [clone: BC96]
Anti-IL2RA Mouse Monoclonal Antibody [clone: BC96]
Catalog # 70-0259-U100
Supplier:  Tonbo Biosciences
undefined
Anti-IL2RA Mouse Monoclonal Antibody [clone: BC96]
Catalog # 70-0259-U100
Supplier:  Tonbo Biosciences

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    Interleukin 2 Receptor Alpha
  • Antigen symbol:
    IL2RA
  • Clonality:
    Monoclonal
  • Clone:
    BC96
  • Conjugation:
    Unconjugated
  • Flow cytometry:
    Yes
  • Host:
    Mouse
  • Isotype:
    IgG1 kappa
  • Reactivity:
    Baboon,
    Cynomolgus monkey,
    Macaque,
    Chimpanzee,
    Human
  • Cross adsorption:
    none
  • Gene ID:
    3559
  • Antigen synonyms:
    TCGFR|IL2-RA|p55|IL-2R subunit alpha|IL-2 receptor subunit alpha|IL-2-RA|IL2R|TAC antigen|IDDM10|CD25
  • Storage temperature:
    Cool
  • Shipping temperature:
    Ice
  • Pk:
    100 µG

Specifications

About this item

The BC96 antibody is specific for human CD25, a 55 kDa surface protein also known as the Interleukin-2 Receptor alpha chain, or IL-2R alpha. CD25 may bind IL-2 by itself, although with low affinity and without induction of cell signaling. CD25 is also expressed within a high-affinity complex, along with the IL-2R beta chain (CD122) and the common gamma chain (CD132), to form a signaling receptor complex. Signaling via CD25 may modulate Th17 and Treg cell differentiation, and has been shown to play a role in the function of dendritic cells.

The BC96 antibody may be used as a marker for expression of CD25 during T and B cell development, on activated mature T and B cells, and for its expression at high levels on natural T regulatory cells (nTreg cells) in the thymus or periphery, and on “induced” peripheral Tregs. This antibody is cross-reactive with CD25 in several non-human species, including Baboon, Chimpanzee, Cynomolgus and Rhesus.

Type: Primary
Antigen: IL2RA (interleukin 2 receptor, alpha)
Clonality: monoclonal
Clone: BC96
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 Kappa
Reactivity: