Rat IgG1 Isotype Control [clone: HRPN] (PerCP (Peridinin-Chlorophyll Protein Complex)-Cy5.5®)
Catalog # 65-4301-U100
Supplier: Tonbo Biosciences
Specifications
- Antibody type:Control
- Clonality:Monoclonal
- Clone:HRPN
- Conjugation:PerCP (Peridinin-Chlorophyll Protein Complex)-Cy5.5®
- Flow cytometry:Yes
- Host:Rat
- Isotype:IgG1
- Cross adsorption:No
- Storage temperature:Store undiluted at 2-8 °C, protect from light, do not freeze.
- Shipping temperature:Ice
- Size:100 μg
- Pk:100 µG
Specifications
About this item
The HRPN immunoglobulin is useful as an isotype-matched control. The HRPN immunolglobulin has an unknown binding specificity and is used as an isotype control for rat IgG1 antibodies.
Type: Control
Antigen: IgG2b
Clonality: Monoclonal
Clone: HRPN
Conjugation: PerCP-Cy5.5®
Epitope:
Host: Rat
Isotype: IgG1
Reactivity: