Order Entry
Czech Republic
ContactUsLinkComponent
Anti-PTPRC Rat Monoclonal Antibody (PE (Phycoerythrin)/Cy7®) [clone: 30-F11]
Anti-PTPRC Rat Monoclonal Antibody (PE (Phycoerythrin)/Cy7®) [clone: 30-F11]
Supplier:  Tonbo Biosciences
Certificates
undefined
Anti-PTPRC Rat Monoclonal Antibody (PE (Phycoerythrin)/Cy7®) [clone: 30-F11]
Supplier:  Tonbo Biosciences

Specifications

  • Catalog No:
  • 60-0451-U100
  • 60-0451-U025
  • Antigen symbol:
  • PTPRC
  • PTPRC
  • Antigen name:
  • Protein tyrosine phosphatase receptor type C
  • Protein tyrosine phosphatase receptor type C
  • Antigen synonyms:
  • CD45R|B220|CD45|GP180|T200|L-CA|LY5|receptor type C|LCA|protein tyrosine phosphatase
  • CD45R|B220|CD45|GP180|T200|L-CA|LY5|receptor type C|LCA|protein tyrosine phosphatase
  • Antibody type:
  • Primary
  • Primary
  • Clonality:
  • Monoclonal
  • Monoclonal
  • Conjugation:
  • PE (Phycoerythrin)/Cy7®
  • PE (Phycoerythrin)/Cy7®
  • Clone:
  • 30-F11
  • 30-F11
  • Reactivity:
  • Mouse
  • Mouse
  • Host:
  • Rat
  • Rat
  • Gene ID:
  • 19264
  • 19264
  • Isotype:
  • IgG2b kappa
  • IgG2b kappa
  • Western blot:
  • Yes
  • Yes
  • ImmunoChemistry:
  • Yes
  • Yes
  • ImmunoPrecipitation:
  • Yes
  • Yes
  • ImmunoFluorescence:
  • Yes
  • Yes
  • Flow cytometry:
  • Yes
  • Yes
  • Cross adsorption:
  • none
  • none
  • Storage temperature:
  • Cool
  • Cool
  • Shipping temperature:
  • Ice
  • Ice

Specifications

Related Information

Frequently Bought Together