Order Entry
Czech Republic
ContactUsLinkComponent
 
Anti-CD44 Rat Monoclonal Antibody (PE (Phycoerythrin)/Cy7®) [clone: IM7]
 
undefined
Anti-CD44 Rat Monoclonal Antibody (PE (Phycoerythrin)/Cy7®) [clone: IM7]

 

  • Catalog No:
  • 60-0441-U025
  • 60-0441-U100
  • Antigen symbol:
  • CD44
  • CD44
  • Antigen name:
  • CD44 antigen
  • CD44 antigen
  • Antigen synonyms:
  • HERMES|AW121933|AW146109|AU023126|Pgp-1|Ly-24
  • HERMES|AW121933|AW146109|AU023126|Pgp-1|Ly-24
  • Antibody type:
  • Primary
  • Primary
  • Clonality:
  • Monoclonal
  • Monoclonal
  • Conjugation:
  • PE (Phycoerythrin)/Cy7®
  • PE (Phycoerythrin)/Cy7®
  • Clone:
  • IM7
  • IM7
  • Reactivity:
  • Baboon, Pig, Horse, Cynomolgus monkey, Macaque, Chimpanzee, Human, Dog, Mouse, Cat
  • Baboon, Pig, Horse, Cynomolgus monkey, Macaque, Chimpanzee, Human, Dog, Mouse, Cat
  • Host:
  • Rat
  • Rat
  • Gene ID:
  • 12505
  • 12505
  • Isotype:
  • IgG2b kappa
  • IgG2b kappa
  • Western blot:
  • Yes
  • Yes
  • ImmunoChemistry:
  • Yes
  • Yes
  • ImmunoPrecipitation:
  • Yes
  • Yes
  • Flow cytometry:
  • Yes
  • Yes
  • Cross adsorption:
  • none
  • none
  • Storage temperature:
  • Cool
  • Cool
  • Shipping temperature:
  • Ice
  • Ice

 

Frequently Bought Together