Order Entry
Czech Republic
ContactUsLinkComponent
Rat IgG2a Isotype Control [clone: 2A3] (PE (Phycoerythrin))
Rat IgG2a Isotype Control [clone: 2A3] (PE (Phycoerythrin))
Catalog # 50-4321-U100
Supplier:  Tonbo Biosciences
Rat IgG2a Isotype Control [clone: 2A3] (PE (Phycoerythrin))
Catalog # 50-4321-U100
Supplier:  Tonbo Biosciences

Specifications

  • Antibody type:
    Control
  • Clonality:
    Monoclonal
  • Clone:
    2A3
  • Conjugation:
    PE (Phycoerythrin)
  • Flow cytometry:
    Yes
  • Host:
    Rat
  • Isotype:
    IgG2a
  • Cross adsorption:
    none
  • Storage temperature:
    Cool
  • Shipping temperature:
    Ice
  • Pk:
    100 µG

Specifications

About this item

The 2A3 immunoglobulin is useful as an isotype-matched control. The 2A3 immunolglobulin has an unknown binding specificity and is used as an isotype control for rat IgG2a antibodies.

Type: Control
Antigen: IgG2a (Rat) (rat IgG2a)
Clonality: monoclonal
Clone: 2A3
Conjugation: PE (Phycoerythrin)
Epitope:
Host: Rat
Isotype: IgG2a
Reactivity: