Order Entry
Czech Republic
ContactUsLinkComponent
Anti-CD45R Rat Monoclonal Antibody (PE (Phycoerythrin)) [clone: RA3-6B2]
Anti-CD45R Rat Monoclonal Antibody (PE (Phycoerythrin)) [clone: RA3-6B2]
Supplier:  Tonbo Biosciences
Certificates
undefined
Anti-CD45R Rat Monoclonal Antibody (PE (Phycoerythrin)) [clone: RA3-6B2]
Supplier:  Tonbo Biosciences

Specifications

  • Catalog No:
  • 50-0452-U025
  • 50-0452-U100
  • Antigen symbol:
  • PTPRC
  • PTPRC
  • Antigen name:
  • Protein tyrosine phosphatase receptor type C
  • Protein tyrosine phosphatase receptor type C
  • Antigen synonyms:
  • CD45R|B220|CD45|GP180|T200|L-CA|LY5|receptor type C|LCA|protein tyrosine phosphatase
  • CD45R|B220|CD45|GP180|T200|L-CA|LY5|receptor type C|LCA|protein tyrosine phosphatase
  • Antibody type:
  • Primary
  • Primary
  • Clonality:
  • Monoclonal
  • Monoclonal
  • Conjugation:
  • PE (Phycoerythrin)
  • PE (Phycoerythrin)
  • Clone:
  • RA3-6B2
  • RA3-6B2
  • Reactivity:
  • Human, Mouse, Cat
  • Human, Mouse, Cat
  • Host:
  • Rat
  • Rat
  • Gene ID:
  • 19264
  • 19264
  • Isotype:
  • IgG2a kappa
  • IgG2a kappa
  • ImmunoChemistry:
  • Yes
  • Yes
  • ImmunoPrecipitation:
  • Yes
  • Yes
  • ImmunoFluorescence:
  • Yes
  • Yes
  • Flow cytometry:
  • Yes
  • Yes
  • Cross adsorption:
  • none
  • none
  • Storage temperature:
  • Cool
  • Cool
  • Shipping temperature:
  • Ice
  • Ice

Specifications

Related Information

Frequently Bought Together