Anti-KLRG1 Mouse Monoclonal Antibody (FITC (Fluorescein)) [clone: 2F1]
Supplier: Tonbo Biosciences
Certificates
About this item
Type: Primary
Antigen: KLRG1 (killer cell lectin-like receptor subfamily G, member 1)
Clonality: monoclonal
Clone: 2F1
Conjugation: FITC (Fluorescein)
Epitope:
Host: Mouse
Isotype: IgG
Reactivity: Mouse
Specifications
- Catalog No:
- 35-5893-U100
- 35-5893-U025
- 35-5893-U500
- Antigen symbol:
- KLRG1
- KLRG1
- KLRG1
- Antigen name:
- Killer cell lectin-like receptor subfamily G member 1
- Killer cell lectin-like receptor subfamily G member 1
- Killer cell lectin-like receptor subfamily G member 1
- Antigen synonyms:
- MAFA|MAFA-2F1|2F1|CLEC15A|MAFA-LIKE|MAFA-L
- MAFA|MAFA-2F1|2F1|CLEC15A|MAFA-LIKE|MAFA-L
- MAFA|MAFA-2F1|2F1|CLEC15A|MAFA-LIKE|MAFA-L
- Antibody type:
- Primary
- Primary
- Clonality:
- Monoclonal
- Monoclonal
- Conjugation:
- FITC (Fluorescein Isothiocyanate)
- FITC (Fluorescein Isothiocyanate)
- FITC (Fluorescein Isothiocyanate)
- Clone:
- 2F1
- 2F1
- 2F1
- Reactivity:
- Mouse
- Mouse
- Mouse
- Host:
- Mouse
- Mouse
- Golden syrian hamster
- Gene ID:
- 50928
- 50928
- 50928
- Isotype:
- IgG
- IgG
- IgG
- ImmunoPrecipitation:
- Yes
- Yes
- Flow cytometry:
- Yes
- Yes
- Yes
- Cross adsorption:
- none
- none
- Storage temperature:
- Cool
- Cool
- Store undiluted at 2-8° C, protect from light, do not freeze.
- Shipping temperature:
- Ice
- Ice
- Tested applications:
- Flow Cytometry
Specifications
Frequently Bought Together