Order Entry
Czech Republic
ContactUsLinkComponent
 
Anti-TNFRSF18 Rat Monoclonal Antibody (FITC (Fluorescein)) [clone: DTA-1]
 
undefined
Anti-TNFRSF18 Rat Monoclonal Antibody (FITC (Fluorescein)) [clone: DTA-1]

 

  • Catalog No:
  • 35-5874-U100
  • 35-5874-U025
  • Antigen symbol:
  • TNFRSF18
  • TNFRSF18
  • Antigen name:
  • Tumor Necrosis Factor Receptor Superfamily Member 18
  • Tumor Necrosis Factor Receptor Superfamily Member 18
  • Antigen synonyms:
  • CD357|TNFRSF18|Activation-inducible TNFR Family Receptor|Glucocorticoid-induced TNFR-related Protein|AITR|Tumor Necrosis Factor Receptor Superfamily Member 18
  • CD357|TNFRSF18|Activation-inducible TNFR Family Receptor|Glucocorticoid-induced TNFR-related Protein|AITR|Tumor Necrosis Factor Receptor Superfamily Member 18
  • Antibody type:
  • Primary
  • Primary
  • Clonality:
  • Monoclonal
  • Monoclonal
  • Conjugation:
  • FITC (Fluorescein Isothiocyanate)
  • FITC (Fluorescein Isothiocyanate)
  • Clone:
  • DTA-1
  • DTA-1
  • Reactivity:
  • Mouse
  • Mouse
  • Host:
  • Rat
  • Rat
  • Gene ID:
  • 21936
  • 21936
  • Isotype:
  • IgG2b
  • IgG2b
  • Flow cytometry:
  • Yes
  • Yes
  • Cross adsorption:
  • none
  • none
  • Storage temperature:
  • Cool
  • Cool
  • Shipping temperature:
  • Ice
  • Ice

 

Frequently Bought Together