About this item
The A7R34 antibody is specific for mouse CD127, a 60 to 90 kDa cell surface protein also known as the Interleukin-7 Receptor alpha chain, or IL-7R alpha. CD127 is typically expressed at the cell surface as a heterodimer with the common gamma chain (CD132). This complex acts as the functional receptor for IL-7, a cytokine important in T and B cell development, and in mature T cell homeostasis. A second cytokine known as Thymic Stromal Lymphopoietin (TSLP) also binds to a receptor complex of CD127 and the TSLPR chain to trigger activation of dendritic cells, and is involved in B cell development, allergy and autoimmunity.
The A7R34 antibody may be used as a phenotypic marker for CD127 on immature B cells, on subsets of thymocytes which are double negative (CD4-CD8- ) or single positive (CD4+ or CD8+), and at low levels on mature, peripheral T cells. CD127 is a key marker, when used in combination with CD4 and CD25, to distinguish Treg and effector/memory Treg populations known as T(REM).
Type: Primary
Antigen: IL7R (interleukin 7 receptor)
Clonality: monoclonal
Clone: A7R34
Conjugation: FITC (Fluorescein)
Epitope:
Host: Rat
Isotype: IgG2a Kappa
Reactivity: Mouse
Specifications
- Catalog No:
- 35-1271-U025
- 35-1271-U100
- 35-1271-U500
- Antigen symbol:
- IL17RA
- IL17RA
- IL17RA
- Antigen name:
- Interleukin 17 Receptor A
- Interleukin 17 Receptor A
- Interleukin 17 Receptor A
- Antigen synonyms:
- IL-17 R|IL-17 RA|Il17r|AW538159|Cdw217|VDw217
- IL-17 R|IL-17 RA|Il17r|AW538159|Cdw217|VDw217
- IL-17 R|IL-17 RA|Il17r|AW538159|Cdw217|VDw217
- Antibody type:
- Primary
- Primary
- Primary
- Clonality:
- Monoclonal
- Monoclonal
- Monoclonal
- Conjugation:
- FITC (Fluorescein Isothiocyanate)
- FITC (Fluorescein Isothiocyanate)
- FITC (Fluorescein Isothiocyanate)
- Clone:
- A7R34
- A7R34
- A7R34
- Reactivity:
- Mouse
- Mouse
- Mouse
- Host:
- Rat
- Rat
- Rat
- Gene ID:
- 16172
- 16172
- 16172
- Isotype:
- IgG2a kappa
- IgG2a kappa
- IgG2a kappa
- Flow cytometry:
- Yes
- Yes
- Yes
- Neutralisation:
- Yes
- Yes
- Yes
- Cross adsorption:
- none
- none
- none
- Storage temperature:
- Store undiluted at 2...8 °C, protect from light, do not freeze
- Store undiluted at 2...8 °C, protect from light, do not freeze
- Store undiluted at 2...8 °C, protect from light, do not freeze
- Shipping temperature:
- Ice
- Ice
- Ice
- Tested applications:
- Flow Cytometry
- Flow Cytometry
- Flow Cytometry