Order Entry
Czech Republic
ContactUsLinkComponent
Anti-CSF1R Rat Monoclonal Antibody (FITC (Fluorescein)) [clone: AFS98]
Anti-CSF1R Rat Monoclonal Antibody (FITC (Fluorescein)) [clone: AFS98]
Supplier:  Tonbo Biosciences
Certificates
undefined
Anti-CSF1R Rat Monoclonal Antibody (FITC (Fluorescein)) [clone: AFS98]
Supplier:  Tonbo Biosciences

Specifications

  • Catalog No:
  • 35-1152-U025
  • 35-1152-U100
  • Antigen symbol:
  • CSF1R
  • CSF1R
  • Antigen name:
  • Colony Stimulating Factor 1 receptor
  • Colony Stimulating Factor 1 receptor
  • Antigen synonyms:
  • CD115|Csfmr|AI323359|Fim-2|M-CSFR|Fms|CSF-1R|M-CSF-R
  • CD115|Csfmr|AI323359|Fim-2|M-CSFR|Fms|CSF-1R|M-CSF-R
  • Antibody type:
  • Primary
  • Primary
  • Clonality:
  • Monoclonal
  • Monoclonal
  • Conjugation:
  • FITC (Fluorescein Isothiocyanate)
  • FITC (Fluorescein Isothiocyanate)
  • Clone:
  • AFS98
  • AFS98
  • Reactivity:
  • Mouse
  • Mouse
  • Host:
  • Rat
  • Rat
  • Gene ID:
  • 12978
  • 12978
  • Isotype:
  • IgG2a kappa
  • IgG2a kappa
  • Western blot:
  • Yes
  • Yes
  • Flow cytometry:
  • Yes
  • Yes
  • Neutralisation:
  • Yes
  • Yes
  • Cross adsorption:
  • none
  • none
  • Storage temperature:
  • Cool
  • Cool
  • Shipping temperature:
  • Ice
  • Ice
  • Tested applications:
  • Flow Cytometry
  • Flow Cytometry

Specifications

Related Information

Frequently Bought Together