The M1/70 antibody reacts with human and mouse CD11b, also known as integrin alpha M. This 165-170 kDa cell surface glycoprotein is part of a family of integrin receptors that mediate adhesion between cells (cell-cell) and components of the extracellular matrix, e.g. fibrinogen (cell-matrix). In addition, integrins are active signaling receptors which recruit leukocytes to inflammatory sites and promote cell activation. Complete, functional integrin receptors consist of distinct combinations of integrin chains which are differentially expressed. Integrin alpha M (CD11b) assembles with Integrin beta-2 (CD18) into a receptor known as Macrophage Antigen-1 (Mac-1) or complement receptor type 3 (CR3). This receptor binds and induces intracellular signaling through ICAM-1 on endothelial cells and can also facilitate removal of iC3b bearing foreign cells.
The M1/70 antibody is widely used as a marker for CD11b expression on mouse macrophages, granulocytes, neutrophils, and NK cells. The antibody is also reported to be cross-reactive for Rhesus macaque CD11b.
Type: Primary
Antigen: ITGAM (integrin, alpha M (complement component 3 receptor 3 subunit))
Clonality: monoclonal
Clone: M1/70
Conjugation: APC (Allophycocyanin)/Cy7®
Epitope:
Host: Rat
Isotype: IgG2b Kappa
Reactivity:
- Catalog No:
- 25-0112-U100
- 25-0112-U025
- Antigen symbol:
- ITGAM
- ITGAM
- Antigen name:
- Integrin Alpha M
- Integrin Alpha M
- Antigen synonyms:
- CD11B|integrin subunit alpha M|MAC-1|MO1A|MAC1A|SLEB6|CR3A
- CD11B|integrin subunit alpha M|MAC-1|MO1A|MAC1A|SLEB6|CR3A
- Antibody type:
- Primary
- Primary
- Clonality:
- Monoclonal
- Monoclonal
- Conjugation:
- APC (Allophycocyanin)/Cy7®
- APC (Allophycocyanin)/Cy7®
- Clone:
- M1/70
- M1/70
- Reactivity:
- Baboon, Cynomolgus monkey, Rhesus monkey, Chimpanzee, Human, Mouse
- Baboon, Cynomolgus monkey, Rhesus monkey, Chimpanzee, Human, Mouse
- Host:
- Rat
- Rat
- Gene ID:
- 16409 / 3684
- 16409 / 3684
- Isotype:
- IgG2b kappa
- IgG2b kappa
- ImmunoChemistry:
- Yes
- Yes
- ImmunoPrecipitation:
- Yes
- Yes
- ImmunoFluorescence:
- Yes
- Yes
- Flow cytometry:
- Yes
- Yes
- Cross adsorption:
- none
- none
- Storage temperature:
- Cool
- Cool
- Shipping temperature:
- Ice
- Ice