Order Entry
Czech Republic
ContactUsLinkComponent
Anti-LRRC32 Mouse Monoclonal Antibody (APC (Allophycocyanin)) [clone: GARP5]
Anti-LRRC32 Mouse Monoclonal Antibody (APC (Allophycocyanin)) [clone: GARP5]
Supplier:  Tonbo Biosciences
Certificates
undefined
Anti-LRRC32 Mouse Monoclonal Antibody (APC (Allophycocyanin)) [clone: GARP5]
Supplier:  Tonbo Biosciences

Specifications

  • Catalog No:
  • 20-9882-T100
  • 20-9882-T025
  • Antigen symbol:
  • LRRC32
  • LRRC32
  • Antigen name:
  • Leucine Rich Repeat Containing 32
  • Leucine Rich Repeat Containing 32
  • Antigen synonyms:
  • GARP|D11S833E
  • GARP|D11S833E
  • Antibody type:
  • Primary
  • Primary
  • Clonality:
  • Monoclonal
  • Monoclonal
  • Conjugation:
  • APC (Allophycocyanin)
  • APC (Allophycocyanin)
  • Clone:
  • GARP5
  • GARP5
  • Reactivity:
  • Human
  • Human
  • Host:
  • Mouse
  • Mouse
  • Gene ID:
  • 2615
  • 2615
  • Isotype:
  • IgG1
  • IgG1
  • Western blot:
  • Yes
  • Yes
  • ImmunoPrecipitation:
  • Yes
  • Yes
  • Flow cytometry:
  • Yes
  • Yes
  • Neutralisation:
  • Yes
  • Yes
  • Cross adsorption:
  • none
  • none
  • Storage temperature:
  • Store undiluted at 2...8 °C, protect from light, do not freeze
  • Store undiluted at 2...8 °C, protect from light, do not freeze
  • Shipping temperature:
  • Ice
  • Ice
  • Tested applications:
  • Flow Cytometry
  • Flow Cytometry

Specifications

Related Information

Frequently Bought Together