Order Entry
Czech Republic
ContactUsLinkComponent
 
Anti-IL4 Rat Monoclonal Antibody (APC (Allophycocyanin)) [clone: 11B11]
 
undefined
Anti-IL4 Rat Monoclonal Antibody (APC (Allophycocyanin)) [clone: 11B11]

 

  • Catalog No:
  • 20-7041-U100
  • 20-7041-U025
  • Antigen symbol:
  • IL4
  • IL4
  • Antigen name:
  • Interleukin 4
  • Interleukin 4
  • Antigen synonyms:
  • BCGF-1|B-cell stimulatory factor 1|IL-4|BCGF1|BSF1|BSF-1|Interleukin-4
  • BCGF-1|B-cell stimulatory factor 1|IL-4|BCGF1|BSF1|BSF-1|Interleukin-4
  • Antibody type:
  • Primary
  • Primary
  • Clonality:
  • Monoclonal
  • Monoclonal
  • Conjugation:
  • APC (Allophycocyanin)
  • APC (Allophycocyanin)
  • Clone:
  • 11B11
  • 11B11
  • Reactivity:
  • Mouse
  • Mouse
  • Host:
  • Rat
  • Rat
  • Gene ID:
  • 16189
  • 16189
  • Isotype:
  • IgG1 kappa
  • IgG1 kappa
  • ELISA:
  • Yes
  • Yes
  • ImmunoChemistry:
  • Yes
  • Yes
  • ImmunoFluorescence:
  • Yes
  • Yes
  • Flow cytometry:
  • Yes
  • Yes
  • Neutralisation:
  • Yes
  • Yes
  • Cross adsorption:
  • none
  • none
  • Storage temperature:
  • Cool
  • Cool
  • Shipping temperature:
  • 4 °C
  • 4 °C
  • Tested applications:
  • Flow Cytometry
  • Flow Cytometry

 

Frequently Bought Together