Order Entry
Czech Republic
ContactUsLinkComponent
Anti-SELL Rat Monoclonal Antibody (APC (Allophycocyanin)) [clone: MEL-14]
Anti-SELL Rat Monoclonal Antibody (APC (Allophycocyanin)) [clone: MEL-14]
Supplier:  Tonbo Biosciences
Certificates
undefined
Anti-SELL Rat Monoclonal Antibody (APC (Allophycocyanin)) [clone: MEL-14]
Supplier:  Tonbo Biosciences

Specifications

  • Catalog No:
  • 20-0621-U100
  • 20-0621-U025
  • Antigen symbol:
  • SELL
  • SELL
  • Antigen name:
  • Selectin L
  • Selectin L
  • Antigen synonyms:
  • LECAM1|CD62L|TQ1|LYAM1|LEU8|LNHR|LSEL|LAM1|PLNHR
  • LECAM1|CD62L|TQ1|LYAM1|LEU8|LNHR|LSEL|LAM1|PLNHR
  • Antibody type:
  • Primary
  • Primary
  • Clonality:
  • Monoclonal
  • Monoclonal
  • Conjugation:
  • APC (Allophycocyanin)
  • APC (Allophycocyanin)
  • Clone:
  • MEL-14
  • MEL-14
  • Reactivity:
  • Mouse
  • Mouse
  • Host:
  • Rat
  • Rat
  • Gene ID:
  • 20343
  • 20343
  • Isotype:
  • IgG2a kappa
  • IgG2a kappa
  • ImmunoChemistry:
  • Yes
  • Yes
  • ImmunoPrecipitation:
  • Yes
  • Yes
  • Flow cytometry:
  • Yes
  • Yes
  • Cross adsorption:
  • none
  • none
  • Storage temperature:
  • Cool
  • Cool
  • Shipping temperature:
  • Ice
  • Ice

Specifications

Related Information

Frequently Bought Together