Anti-KIR2DS2 Rabbit Polyclonal Antibody
Catalog # ABGEAP5764C
Supplier: Abgent
Specifications
- Antibody type:Primary
- Antigen name:Killer cell immunoglobulin-like Receptor two Domains short cytoplasmic tail 2
- Antigen symbol:KIR2DS2
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- Reactivity:Human
- Western blot:Yes
- Epitope:middle
- Antigen synonyms:Serine/threonine-protein kinase PCTAIRE-1|PCTK1|cyclin-dependent kinase 16|CDK16
- Storage temperature:Cool
- Shipping temperature:Ice
- Pk:400 µl
Specifications
About this item
Type: Primary
Antigen: KIR2DS2 (killer cell immunoglobulin-like receptor, two domains, short cytoplasmic tail, 2)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: middle
Host: Rabbit
Isotype: none
Reactivity: Human