Order Entry
Czech Republic
ContactUsLinkComponent
Anti-M6PR Rabbit Polyclonal Antibody
Anti-M6PR Rabbit Polyclonal Antibody
Catalog # ABGEAP2792A
Supplier:  Abgent
undefined
Anti-M6PR Rabbit Polyclonal Antibody
Catalog # ABGEAP2792A
Supplier:  Abgent

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    Mannose-6-phosphate receptor
  • Antigen symbol:
    M6PR
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • ELISA:
    Yes
  • Host:
    Rabbit
  • ImmunoChemistry:
    Yes
  • Reactivity:
    Human,
    Mouse
  • Western blot:
    Yes
  • Epitope:
    N-terminal
  • Antigen synonyms:
    MPR46|MPRD|M6PR|MPR 46|Cation-dependent mannose-6-phosphate receptor|CD Man-6-P receptor|46 kDa mannose 6-phosphate receptor|CD-MPR
  • Storage temperature:
    Cool
  • Shipping temperature:
    Ice
  • Pk:
    400 µl

Specifications

About this item

Type: Primary
Antigen: M6PR (mannose-6-phosphate receptor (cation dependent))
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: N-terminal
Host: Rabbit
Isotype: none
Reactivity: Human, Mouse