Anti-EIF4E2 Rabbit Polyclonal Antibody
Catalog # ABGEAP1955D
Supplier: Abgent
Jump to:
Product Details & Documents
Type: Primary
Antigen: EIF4E2 (eukaryotic translation initiation factor 4E family member 2)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human
Documents
AP1955d.pdf
Technický list
Specifications
- Western blot:Yes
- Clonality:Polyclonal
- Antibody type:Primary
- Storage temperature:Cool
- Host:Rabbit
- Shipping temperature:Ice
- Antigen name:Raf-1 Proto-Oncogene, Serine/Threonine Kinase
- Cat. No.:ABGEAP1955D
- Reactivity:Human
- Antigen synonyms:mRNA cap-binding protein 4EHP|EIF4E2|mRNA cap-binding protein type 3|eIF-4E type 2|eIF4E-like protein 4E-LP|EIF4EL3|Eukaryotic translation initiation factor 4E type 2|Eukaryotic translation initiation factor 4E-like 3|eIF4E type 2|Eukaryotic translation initiation factor 4E homologous protein
- ImmunoChemistry:Yes
- Conjugation:Unconjugated
- Antigen symbol:EIF4E2
- ELISA:Yes



