Anti-PI3KC3 Rabbit Polyclonal Antibody
Catalog # ABGEAP1851A
Supplier: Abgent
Specifications
- Antibody type:Primary
- Antigen name:Phosphatidylinositol-4-Phosphate 3-Kinase, Catalytic Subunit Type 3
- Antigen symbol:PIK3C3
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- Reactivity:Human
- Western blot:Yes
- Epitope:N-terminal
- Antigen synonyms:Phosphatidylinositol 3-kinase catalytic subunit type 3|Phosphatidylinositol 3-kinase p100 subunit|PI3-kinase type 3|PIK3C3|hVps34|VPS34
- Sequence:G24
- Storage temperature:Cool
- Shipping temperature:Ice
- Pk:400 µl
Specifications
About this item
Type: Primary
Antigen: PI3KC3
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: N-terminal
Host: Rabbit
Isotype: none
Reactivity: Human