Order Entry
Czech Republic
ContactUsLinkComponent
Anti-AES Rabbit Polyclonal Antibody
Anti-AES Rabbit Polyclonal Antibody
  ABGEAP14115B
 :  Abgent
Anti-AES Rabbit Polyclonal Antibody
  ABGEAP14115B
 :  Abgent

 

  • Antibody type:
    Primary
  • Antigen name:
    Amino-terminal Enhancer of Split
  • Antigen symbol:
    AES
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • ELISA:
    Yes
  • Host:
    Rabbit
  • Reactivity:
    Human
  • Western blot:
    Yes
  • Epitope:
    C-terminal
  • Antigen synonyms:
    AES-1|AES-2|TLE5|Grg-5|ESP1|GRG5|GRG
  • Storage temperature:
    Cool
  • Shipping temperature:
    Ice
  • Pk:
    400 µl

 

 

Type: Primary
Antigen: AES (amino-terminal enhancer of split)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: C-terminal
Host: Rabbit
Isotype: none
Reactivity: Human