Anti-ARPP21 Rabbit Polyclonal Antibody
ABGEAP13040A
: Abgent
- Antibody type:Primary
- Antigen name:cAMP-regulated phosphoprotein 21kDa
- Antigen symbol:ARPP-21
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- Reactivity:Human
- Western blot:Yes
- Epitope:N-terminal
- Antigen synonyms:RCS|TARPP|R3HDM3
- Storage temperature:Cool
- Shipping temperature:Ice
- Pk:400 µl
Type: Primary
Antigen: ARPP21 (cAMP-regulated phosphoprotein, 21kDa)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: N-terminal
Host: Rabbit
Isotype: none
Reactivity: Human