Anti-KATNBL1 Rabbit Polyclonal Antibody
Catalog # AVIVARP68900_P050
Supplier: Aviva Systems Biology
Specifications
- Antibody type:Primary
- Antigen name:katanin p80 subunit B-like 1
- Antigen symbol:KATNBL1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Reactivity:Human
- Western blot:Yes
- Epitope:N-terminal
- Cross adsorption:none
- Gene ID:79768
- Antigen synonyms:C15orf29
- Storage temperature:Cool
- Shipping temperature:Ice
- Pk:50 µG
Specifications
About this item
Type: Primary
Antigen: KATNBL1 (katanin p80 subunit B-like 1)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: N-terminal
Host: Rabbit
Isotype: none
Reactivity: Human