Anti-NOL9 Rabbit Polyclonal Antibody
Catalog # AVIVARP68888_P050
Supplier: Aviva Systems Biology
Specifications
- Antibody type:Primary
- Antigen name:Nucleolar protein 9
- Antigen symbol:NOL9
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Reactivity:Human
- Western blot:Yes
- Epitope:N-terminal
- Cross adsorption:none
- Gene ID:79707
- Antigen synonyms:Grc3|NET6
- Storage temperature:Cool
- Shipping temperature:Ice
- Pk:50 µG
Specifications
About this item
Type: Primary
Antigen: NOL9 (nucleolar protein 9)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: N-terminal
Host: Rabbit
Isotype: none
Reactivity: Human