Anti-CPSF3L Rabbit Polyclonal Antibody
Katalogové číslo AVIVARP68235_P050
Dodavatel: Aviva Systems Biology
Specifikace
- Antibody type:Primary
- Antigen name:cleavage and polyadenylation specific factor 3-like
- Antigen symbol:CPSF3L
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Reactivity:Human
- Western blot:Yes
- Epitope:C-terminal
- Cross adsorption:none
- Storage temperature:Cool
- Shipping temperature:Ice
- Pk:50 µG
Specifikace
O tomto produktu
Type: Primary
Antigen: CPSF3L (cleavage and polyadenylation specific factor 3-like)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: C-terminal
Host: Rabbit
Isotype: none
Reactivity: Human