Anti-CLEC4F Rabbit Polyclonal Antibody
Catalog # AVIVARP68220_P050
Supplier: Aviva Systems Biology
Specifications
- Antibody type:Primary
- Antigen name:C-type lectin domain family 4, member F
- Antigen symbol:CLEC4F
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Reactivity:Human
- Western blot:Yes
- Epitope:C-terminal
- Cross adsorption:none
- Gene ID:165530
- Antigen synonyms:KCLR|CLECSF13
- Storage temperature:Cool
- Shipping temperature:Ice
- Pk:50 µG
Specifications
About this item
Type: Primary
Antigen: CLEC4F (C-type lectin domain family 4, member F)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: C-terminal
Host: Rabbit
Isotype: none
Reactivity: Human