Anti-CBWD2 Rabbit Polyclonal Antibody
AVIVARP68167_P050
- Antibody type:Primary
- Antigen name:COBW domain containing 2
- Antigen symbol:CBWD2
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Reactivity:Human
- Western blot:Yes
- Epitope:C-terminal
- Cross adsorption:none
- Gene ID:150472
- Storage temperature:Cool
- Shipping temperature:Ice
- Pk:50 µG
Type: Primary
Antigen: CBWD2 (COBW domain containing 2)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: C-terminal
Host: Rabbit
Isotype: none
Reactivity: Human