Anti-C11orf2 Rabbit Polyclonal Antibody
AVIVARP67416_P050
- Antibody type:Primary
- Antigen name:vacuolar protein sorting 51 homolog (S. cerevisiae)
- Antigen symbol:VPS51
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Reactivity:Human
- Western blot:Yes
- Epitope:C-terminal
- Cross adsorption:none
- Gene ID:738
- Antigen synonyms:C11orf2|ANG2|FFR|ANG3|C11orf3
- Storage temperature:Cool
- Shipping temperature:Ice
- Pk:50 µG
Type: Primary
Antigen: C11orf2
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: C-terminal
Host: Rabbit
Isotype: none
Reactivity: Human