Anti-MINPP1 Rabbit Polyclonal Antibody
AVIVARP66212_P050
- Antibody type:Primary
- Antigen name:multiple inositol-polyphosphate phosphatase 1
- Antigen symbol:MINPP1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Reactivity:Human
- Western blot:Yes
- Epitope:C-terminal
- Cross adsorption:none
- Gene ID:9562
- Antigen synonyms:HIPER1|MINPP2|MIPP
- Storage temperature:Cool
- Shipping temperature:Ice
- Pk:50 µG
Type: Primary
Antigen: MINPP1 (multiple inositol-polyphosphate phosphatase 1)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: C-terminal
Host: Rabbit
Isotype: none
Reactivity: Human