Order Entry
Czech Republic
ContactUsLinkComponent
Anti-LILRB5 Rabbit Polyclonal Antibody
Anti-LILRB5 Rabbit Polyclonal Antibody
Catalog # AVIVARP63625_P050
undefined
Anti-LILRB5 Rabbit Polyclonal Antibody
Catalog # AVIVARP63625_P050

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    leukocyte immunoglobulin-like receptor, subfamily B (with TM and ITIM domains), member 5
  • Antigen symbol:
    LILRB5
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • Host:
    Rabbit
  • Reactivity:
    Human
  • Western blot:
    Yes
  • Epitope:
    C-terminal
  • Cross adsorption:
    none
  • Gene ID:
    10990
  • Antigen synonyms:
    LIR-8|LIR8|CD85C
  • Storage temperature:
    Cool
  • Shipping temperature:
    Ice
  • Pk:
    50 µG

Specifications

About this item

Type: Primary
Antigen: LILRB5 (leukocyte immunoglobulin-like receptor, subfamily B (with TM and ITIM domains), member 5)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: C-terminal
Host: Rabbit
Isotype: none
Reactivity: Human