Anti-CRHR1 Rabbit Polyclonal Antibody
Catalog # AVIVARP63607_P050
Supplier: Aviva Systems Biology
Specifications
- Antibody type:Primary
- Antigen name:Corticotropin releasing hormone receptor 1
- Antigen symbol:CRHR1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Western blot:Yes
- Epitope:C-terminal
- Cross adsorption:none
- Gene ID:1394
- Antigen synonyms:CRH-R1|CRH-R-1|CRF1|CRF-R|CRHR|CRF-R1|CRFR1|CRHR1L|CRFR-1|CRF-R-1
- Storage temperature:Cool
- Shipping temperature:Ice
- Pk:50 µG
Specifications
About this item
Type: Primary
Antigen: CRHR1 (corticotropin releasing hormone receptor 1)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: C-terminal
Host: Rabbit
Isotype: none
Reactivity: