Anti-HTRA1 Rabbit Polyclonal Antibody
Catalog # AVIVARP63548_P050
Supplier: Aviva Systems Biology
Specifications
- Antibody type:Primary
- Antigen name:HtrA serine peptidase 1
- Antigen symbol:HTRA1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Reactivity:Pig,Human,Bovine,Rat,Mouse
- Western blot:Yes
- Epitope:N-terminal
- Cross adsorption:none
- Gene ID:5654
- Antigen synonyms:ARMD7|HtrA|ORF480|PRSS11|CARASIL|L56
- Storage temperature:Cool
- Shipping temperature:Ice
- Pk:50 µG
Specifications
About this item
Type: Primary
Antigen: HTRA1 (HtrA serine peptidase 1)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: N-terminal
Host: Rabbit
Isotype: none
Reactivity: Bovine, Human, Mouse, Pig, Rat