Anti-DLG3 Rabbit Polyclonal Antibody
Catalog # AVIVARP63505_P050
Supplier: Aviva Systems Biology
Specifications
- Antibody type:Primary
- Antigen name:Discs, large homolog 3 (Drosophila)
- Antigen symbol:DLG3
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Western blot:Yes
- Epitope:middle
- Cross adsorption:none
- Gene ID:53310
- Antigen synonyms:SAP102|mKIAA1232|Dlgh3
- Storage temperature:Cool
- Shipping temperature:Ice
- Pk:50 µG
Specifications
About this item
Type: Primary
Antigen: DLG3 (discs, large homolog 3 (Drosophila))
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: middle
Host: Rabbit
Isotype: none
Reactivity: