Anti-CTSH Rabbit Polyclonal Antibody
AVIVARP63370_P050
- Antibody type:Primary
- Antigen name:cathepsin H
- Antigen symbol:CTSH
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Western blot:Yes
- Epitope:C-terminal
- Cross adsorption:none
- Gene ID:1512
- Antigen synonyms:ACC5|minichain|ACC4|CPSB|ACC-4|ACC-5
- Storage temperature:Cool
- Shipping temperature:Ice
- Pk:50 µG
Type: Primary
Antigen: CTSH (cathepsin H)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: C-terminal
Host: Rabbit
Isotype: none
Reactivity: