Anti-TES Rabbit Polyclonal Antibody
Catalog # AVIVARP63068_P050
Supplier: Aviva Systems Biology
Specifications
- Antibody type:Primary
- Antigen name:testin LIM domain protein
- Antigen symbol:TES
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Western blot:Yes
- Epitope:middle
- Cross adsorption:none
- Gene ID:26136
- Antigen synonyms:TESS|TESS-2
- Storage temperature:Cool
- Shipping temperature:Ice
- Pk:50 µG
Specifications
About this item
Type: Primary
Antigen: TES (testis derived transcript (3 LIM domains))
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: middle
Host: Rabbit
Isotype: none
Reactivity: